missing translation for 'onlineSavingsMsg'
Learn More

Maltose, Monohydrate, NF, 92%, Spectrum™ Chemical

Catalog No. p-7157699
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604172 500 g Poly Bottle
18604173 2.5 kg Poly Pail
18604174 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18604172 Supplier Spectrum Chemical Mfg Cor Supplier No. M1079500GM
Only null left

M1079, 6363-53-7, C12H22O11·H2O

Spectrum™ Chemical Maltose, Monohydrate, NF is used in pharmaceutical formulations and as a parenteral supplement of sugar for diabetics. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6363-53-7
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.31
MDL Number MFCD00149343
InChI Key HBDJFVFTHLOSDW-UHFFFAOYNA-N
IUPAC Name 2,3,5,6-tetrahydroxy-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanal hydrate
SMILES O.OCC(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)C(O)C=O

Specifications

CAS Max % 100%
Ignition Residue 0.05%
Packaging Poly Bottle
Quantity 500 g
Percent Purity 92%
Grade NF