missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Maltol, FCC, 99%, Spectrum™ Chemical
M1189, 118-71-8, C6H6O3
$246.09 - $9682.06
Chemical Identifiers
| CAS | 118-71-8 |
|---|---|
| Molecular Formula | C6H6O3 |
| Molecular Weight (g/mol) | 126.11 |
| MDL Number | MFCD00006578 |
| InChI Key | XPCTZQVDEJYUGT-UHFFFAOYSA-N |
| IUPAC Name | 3-hydroxy-2-methyl-4H-pyran-4-one |
| SMILES | CC1=C(O)C(=O)C=CO1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18604168
|
Spectrum Chemical Mfg Cor
M118925GM |
25 g | Amber Glass Bottle |
Each for $246.09
|
|
||||
|
18604169
|
Spectrum Chemical Mfg Cor
M1189100GM |
100 g | Amber Glass Bottle |
Each for $789.83
|
|
||||
|
18604170
|
Spectrum Chemical Mfg Cor
M1189500GM |
500 g | Amber Glass Bottle |
Each for $2,727.89
|
|
||||
|
18604171
|
Spectrum Chemical Mfg Cor
M118925KGBL |
25 kg | Fiber Drum |
Each for $9,682.06
|
|
||||
Description
Spectrum™ Chemical Maltol, FCC is used as a flavor enhancer in breads and cakes. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 118-71-8 | |
| 126.11 | |
| XPCTZQVDEJYUGT-UHFFFAOYSA-N | |
| CC1=C(O)C(=O)C=CO1 |
| C6H6O3 | |
| MFCD00006578 | |
| 3-hydroxy-2-methyl-4H-pyran-4-one |
Specifications
| 118-71-8 | |
| 0.2% | |
| Amber Glass Bottle | |
| MFCD00006578 | |
| XPCTZQVDEJYUGT-UHFFFAOYSA-N | |
| 3-hydroxy-2-methyl-4H-pyran-4-one | |
| 99% |
| 100% | |
| 160-164 C | |
| C6H6O3 | |
| 25 g | |
| CC1=C(O)C(=O)C=CO1 | |
| 126.11 | |
| FCC |