missing translation for 'onlineSavingsMsg'
Learn More

Maltol, FCC, 99%, Spectrum™ Chemical

Catalog No. 18604171
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
25 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604171 25 kg Amber Glass Bottle
18604168 25 g Amber Glass Bottle
18604169 100 g Amber Glass Bottle
18604170 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18604171 Supplier Spectrum Chemical Mfg Cor Supplier No. M118925KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

M1189, 118-71-8, C6H6O3

Spectrum™ Chemical Maltol, FCC is used as a flavor enhancer in breads and cakes. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 118-71-8
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
MDL Number MFCD00006578
InChI Key XPCTZQVDEJYUGT-UHFFFAOYSA-N
IUPAC Name 3-hydroxy-2-methyl-4H-pyran-4-one
SMILES CC1=C(O)C(=O)C=CO1

Specifications

CAS Max % 100%
Ignition Residue 0.2%
Melting Range 160°C to 164°C
Packaging Amber Glass Bottle
Quantity 25 kg
Percent Purity 99%
Grade FCC
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Maltol