missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Maltitol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 151°C |
| Color | White |
| Quantity | 500 g |
| Formula Weight | 344.31 |
| Percent Purity | ≥98.0% (GC) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Maltitol |
Safety and Handling
| EINECSNumber | (8)-0065 |
| RTECSNumber | LZ4394000 |
| TSCA | Yes |