missing translation for 'onlineSavingsMsg'
Learn More

Maltitol 95.0+%, TCI America™

Catalog No. M060125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M060125G 25 g
M0601500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M060125G Supplier TCI America Supplier No. M060125G

Chemical Identifiers

CAS 585-88-6
Molecular Formula C12H24O11
Molecular Weight (g/mol) 344.313
MDL Number MFCD00006600
InChI Key VQHSOMBJVWLPSR-WUJBLJFYSA-N
Synonym maltitol, d-maltitol, maltisorb, malbit, amalti syrup, malti mr, amalty mr 100, amalty, 4-o-alpha-d-glucopyranosyl-d-glucitol, unii-d65dg142wk
PubChem CID 493591
ChEBI CHEBI:68428
IUPAC Name (2S,3R,4R,5R)-4-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexane-1,2,3,5,6-pentol
SMILES C(C1C(C(C(C(O1)OC(C(CO)O)C(C(CO)O)O)O)O)O)O

Specifications

Melting Point 146°C
Color White
Quantity 25 g
Formula Weight 344.31
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Maltitol
EINECSNumber (8)-0065
RTECSNumber LZ4394000
TSCA Yes