missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Maltitol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 585-88-6 |
| Physical Form | Neat |
| Linear Formula | C12H24O11 |
| MDL Number | MFCD00006600 |
| Melting Point | 149°C to 152°C |
| Synonym | 4-O-alpha-Glucopyranosyl-D-sorbitol |
| Type | Certified Reference Material |
| Quantity | 500 mg |
| Formula Weight | 344.31 |
| Grade | Certified Reference Material |