missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Maltitol, 95%

Catalog No. AC295800250
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC295800250 25 g Glass bottle
AC295802500 250 g Glass bottle
AC295800010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC295800250 Supplier Thermo Scientific Chemicals Supplier No. 295800250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 585-88-6 | C12H24O11 | 344.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 585-88-6
Molecular Formula C12H24O11
Molecular Weight (g/mol) 344.32
MDL Number MFCD00006600
InChI Key VQHSOMBJVWLPSR-WUJBLJFYSA-N
Synonym maltitol, d-maltitol, maltisorb, malbit, amalti syrup, malti mr, amalty mr 100, amalty, 4-o-alpha-d-glucopyranosyl-d-glucitol, unii-d65dg142wk
PubChem CID 493591
ChEBI CHEBI:68428
IUPAC Name (2S,3R,4R,5R)-4-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexane-1,2,3,5,6-pentol
SMILES C(C1C(C(C(C(O1)OC(C(CO)O)C(C(CO)O)O)O)O)O)O

Specifications

Melting Point 147°C to 153°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 17, V,7, 145
Specific Rotation Condition + 107.00 (20.00°C c=5,water)
Specific Rotation + 107
Solubility Information (10% in water) Clear colorless
Formula Weight 344.32
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Maltitol, 95%
Show More Show Less
EINECSNumber 209-567-0
RTECSNumber LZ4394000
TSCA TSCA

RUO – Research Use Only