missing translation for 'onlineSavingsMsg'
Learn More

Malonyl dichloride, 94%

Catalog No. AAH5986822
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5986822 100 g
AAH5986814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5986822 Supplier Thermo Scientific Chemicals Supplier No. H5986822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1663-67-8 | C3H2Cl2O2 | 140.947 g/mol

Malonyl Dichloride is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Malonyl Dichloride is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.

Solubility
Decomposes in water.

Notes
Recommended storage temperature: 2 - 4°C. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1663-67-8
Molecular Formula C3H2Cl2O2
Molecular Weight (g/mol) 140.947
MDL Number MFCD00000735
InChI Key SXYFKXOFMCIXQW-UHFFFAOYSA-N
Synonym malonyl chloride, malonyl dichloride, malonoyl chloride, malonic acid dichloride, malonoyl dichloride, malonic acid chloride, malonylchloride, methane-1,1-dicarbonyl chloride, malonylchlorid, malonyldichloride
PubChem CID 74269
IUPAC Name propanedioyl dichloride
SMILES C(C(=O)Cl)C(=O)Cl

Specifications

Density 1.449
Boiling Point 53°C to 55°C (19 mmHg)
Flash Point 47°C (117°F)
Refractive Index 1.465
Linear Formula CH2(COCl)2
Quantity 100 g
UN Number UN2920
Beilstein 774044
Solubility Information Decomposes in water.
Formula Weight 140.95
Percent Purity 97%
Chemical Name or Material Malonyl dichloride
Show More Show Less
Hazard Category H226-H314
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 216-772-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only