accessibility menu, dialog, popup

UserName

ClOClO

CAS RN 1663-67-8

IUPAC Name: propanedioyl dichloride
Synonyms: propanedioyl dichloride
Molecular Weight (g/mol): 140.95
Molecular Formula: C3H2Cl2O2
InChi Key: SXYFKXOFMCIXQW-UHFFFAOYSA-N
SMILES: ClC(=O)CC(Cl)=O
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)