Learn More
Malononitrile, 99%
CAS: 109-77-3 | C3H2N2 | 66.063 g/mol
Supplier: Thermo Scientific Chemicals A150460E
Description
Crosslinking agent for modification of proteins via amidationMalononitrile is used as a precursor in the preparation of anti-cancer drug, triamterene, adenine, methotrexate, thiamine, acrylic fibers and dyes. It is used in Knoevenagel condensation to prepare 2-chlorobenzalmalononitrile by reacting with 2-chlorobenzaldehyde. It acts as a leaching agent for gold. It is used as a raw material for the Gewald reaction of ketone or aldehyde to produce 2-aminothiophene using elemental sulfur and a base. Further, it plays an important role in the preparation of [1,6]-naphthyridines, gamma-ketoamides, 1,4-dihydropyridine, chromeno[2,3-b]pyridine and dihydro-1,4-dithiepine frameworks.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCrosslinking agent for modification of proteins via amidationMalononitrile is used as a precursor in the preparation of anti-cancer drug, triamterene, adenine, methotrexate, thiamine, acrylic fibers and dyes. It is used in Knoevenagel condensation to prepare 2-chlorobenzalmalononitrile by reacting with 2-chlorobenzaldehyde. It acts as a leaching agent for gold. It is used as a raw material for the Gewald reaction of ketone or aldehyde to produce 2-aminothiophene using elemental sulfur and a base. Further, it plays an important role in the preparation of [1,6]-naphthyridines, gamma-ketoamides, 1,4-dihydropyridine, chromeno[2,3-b]pyridine and dihydro-1,4-dithiepine frameworks.
Solubility
Soluble in water, acetone, acetic acid, chloroform, ethanol, ether and benzene.
Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong reducing agents, strong acids and strong bases.
Specifications
| Malononitrile | |
| 30°C to 34°C | |
| 220°C to 222°C | |
| C3H2N2 | |
| MFCD00001883 | |
| UN2647 | |
| 14,5711 | |
| Soluble in water,acetone,acetic acid,chloroform,ethanol,ether and benzene. | |
| C(C#N)C#N | |
| 66.063 | |
| CHEBI:33186 | |
| 99% |
| 109-77-3 | |
| 1.049 | |
| 112°C (233°F) | |
| 1.415 | |
| 2500 g | |
| 773697 | |
| malononitrile, dicyanomethane, cyanoacetonitrile, malonic dinitrile, methylene cyanide, dicyanmethane, malonodinitrile, malonic acid dinitrile, methylenedinitrile, methane, dicyano | |
| CUONGYYJJVDODC-UHFFFAOYSA-N | |
| propanedinitrile | |
| 8010 | |
| 66.06 |
Chemical Identifiers
| 109-77-3 | |
| MFCD00001883 | |
| malononitrile, dicyanomethane, cyanoacetonitrile, malonic dinitrile, methylene cyanide, dicyanmethane, malonodinitrile, malonic acid dinitrile, methylenedinitrile, methane, dicyano | |
| CHEBI:33186 |
| C3H2N2 | |
| CUONGYYJJVDODC-UHFFFAOYSA-N | |
| 8010 | |
| propanedinitrile |
Safety and Handling
GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
H301+H311+H331
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MALONONITRILE
EINECSNumber : 203-703-2
RTECSNumber : OO3150000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only