missing translation for 'onlineSavingsMsg'
Learn More

Malononitrile, 99%

Catalog No. p-7022325
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
2500 g
500 g
Catalog No. Quantity
AAA1504622 100 g
AAA1504636 500 g
AAA150460E 2500 g
3 options

Catalog No. AAA1504622

Supplier: Thermo Scientific Chemicals A1504622

Only null left

CAS: 109-77-3 | C3H2N2 | 66.063 g/mol

Crosslinking agent for modification of proteins via amidationMalononitrile is used as a precursor in the preparation of anti-cancer drug, triamterene, adenine, methotrexate, thiamine, acrylic fibers and dyes. It is used in Knoevenagel condensation to prepare 2-chlorobenzalmalononitrile by reacting with 2-chlorobenzaldehyde. It acts as a leaching agent for gold. It is used as a raw material for the Gewald reaction of ketone or aldehyde to produce 2-aminothiophene using elemental sulfur and a base. Further, it plays an important role in the preparation of [1,6]-naphthyridines, gamma-ketoamides, 1,4-dihydropyridine, chromeno[2,3-b]pyridine and dihydro-1,4-dithiepine frameworks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Crosslinking agent for modification of proteins via amidationMalononitrile is used as a precursor in the preparation of anti-cancer drug, triamterene, adenine, methotrexate, thiamine, acrylic fibers and dyes. It is used in Knoevenagel condensation to prepare 2-chlorobenzalmalononitrile by reacting with 2-chlorobenzaldehyde. It acts as a leaching agent for gold. It is used as a raw material for the Gewald reaction of ketone or aldehyde to produce 2-aminothiophene using elemental sulfur and a base. Further, it plays an important role in the preparation of [1,6]-naphthyridines, gamma-ketoamides, 1,4-dihydropyridine, chromeno[2,3-b]pyridine and dihydro-1,4-dithiepine frameworks.

Solubility
Soluble in water, acetone, acetic acid, chloroform, ethanol, ether and benzene.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong reducing agents, strong acids and strong bases.

Chemical Identifiers

CAS 109-77-3
Molecular Formula C3H2N2
MDL Number MFCD00001883
InChI Key CUONGYYJJVDODC-UHFFFAOYSA-N
Synonym malononitrile, dicyanomethane, cyanoacetonitrile, malonic dinitrile, methylene cyanide, dicyanmethane, malonodinitrile, malonic acid dinitrile, methylenedinitrile, methane, dicyano
PubChem CID 8010
ChEBI CHEBI:33186
IUPAC Name propanedinitrile

Specifications

Melting Point 30°C to 34°C
Density 1.049
Boiling Point 220°C to 222°C
Flash Point 112°C (233°F)
Refractive Index 1.415
Quantity 100 g
UN Number UN2647
Beilstein 773697
Merck Index 14,5711
Solubility Information Soluble in water,acetone,acetic acid,chloroform,ethanol,ether and benzene.
SMILES C(C#N)C#N
Molecular Weight (g/mol) 66.063
Formula Weight 66.06
Percent Purity 99%
Chemical Name or Material Malononitrile
Show More Show Less
Hazard Category H301+H311+H331
Hazard Statement GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MALONONITRILE
EINECSNumber 203-703-2
RTECSNumber OO3150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only