missing translation for 'onlineSavingsMsg'
Learn More

Malonic Acid, TCI America™

Catalog No. p-7131937
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M002825G 25 g
M0028500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M002825G Supplier TCI America Supplier No. M002825G
Only null left

Chemical Identifiers

CAS 141-82-2
Molecular Formula C3H4O4
Molecular Weight (g/mol) 104.061
MDL Number MFCD00002707
InChI Key OFOBLEOULBTSOW-UHFFFAOYSA-N
Synonym malonic acid, dicarboxymethane, carboxyacetic acid, methanedicarboxylic acid, kyselina malonova, usaf ek-695, dicarboxylate, dicarboxylic acid, kyselina malonova czech, propanediolic acid
PubChem CID 867
ChEBI CHEBI:30794
IUPAC Name propanedioic acid
SMILES C(C(=O)O)C(=O)O

Specifications

Melting Point 134°C
Color White
Quantity 25 g
Formula Weight 104.06
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Malonic Acid
EINECSNumber (2)-0912
RTECSNumber OO0175000
TSCA Yes