Learn More
Malonic acid, 99%
Description
Malonic acid is acts as a building block in organic synthesis. It is also useful as a precursor for polyesters and alkyd resins, which is used in coating applications, thereby protecting against UV light, corrosion and oxidation. It acts as a cross linker in the coating industry and surgical adhesive. It finds application in the production of specialty chemicals, flavors and fragrances, polymer cross linkers and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Malonic acid is acts as a building block in organic synthesis. It is also useful as a precursor for polyesters and alkyd resins, which is used in coating applications, thereby protecting against UV light, corrosion and oxidation. It acts as a cross linker in the coating industry and surgical adhesive. It finds application in the production of specialty chemicals, flavors and fragrances, polymer cross linkers and pharmaceuticals.
Solubility
Soluble in cold water.
Notes
Incompatible with bases, oxidizing agents and reducing agents.
Specifications
Specifications
| Melting Point | 132°C to 136°C (decomposition) |
| Density | 1.62 |
| Flash Point | 157°C (314°F) |
| Quantity | 100 g |
| Beilstein | 1751370 |
| Merck Index | 14,5710 |
| Solubility Information | Soluble in cold water. |
| Formula Weight | 104.06 |
| Percent Purity | 99% |
| Chemical Name or Material | Malonic acid |
Safety and Handling
| Hazard Category | H302+H332-H315-H318-H335 |
| Hazard Statement | GHS H Statement H318-H302-H335-H315 Causes serious eye damage. Harmful if swallowed. May cause respiratory irritation. Causes skin irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 205-503-0 |
| RTECSNumber | OO0175000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only