missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Malondialdehyde tetrabutylammonium salt, ≥97.0% (NT), MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 100683-54-3 |
| Linear Formula | OCHCH=CHO[N(CH2 CH2 CH2 CH3)4 ] |
| MDL Number | MFCD00043410 |
| Melting Point | 126°C to 132°C |
| Synonym | Malonaldehyde tetrabutylammonium salt; Tetrabutylammonium malondialdehyde enolate |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 313.52 |
| Percent Purity | ≥97.0% (NT) |
| Grade | Analytical Standard |