missing translation for 'onlineSavingsMsg'
Learn More

Malonamamidine hydrochloride, 98+%

Catalog No. AAB2556914
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2556914 25 g
AAB2556922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2556914 Supplier Thermo Scientific Chemicals Supplier No. B2556914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 34570-17-7 | C3H7ClN3O | 136.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34570-17-7
Molecular Formula C3H7ClN3O
Molecular Weight (g/mol) 136.56
MDL Number MFCD00013007
InChI Key MDSJPVJGEXMXIC-UHFFFAOYSA-N
Synonym malonamamidine hydrochloride, 3-amino-3-iminopropanamide hydrochloride, amidinomalonamide hydrochloride, 2-carbamimidoylacetamide hydrochloride, propanamide, 3-amino-3-imino-, monohydrochloride, amidinomalonamide hcl, amindinomalonamide, malonamideamidine hydrochloride, tos-bb-1115
PubChem CID 3084595
IUPAC Name 3-amino-3-iminopropanamide;hydrochloride
SMILES [Cl].NC(=N)CC(N)=O

Specifications

Melting Point 170°C to 174°C
Quantity 25 g
Formula Weight 137.57
Percent Purity ≥98%
Chemical Name or Material Malonamamidine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 252-095-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only