accessibility menu, dialog, popup

UserName

CAS RN 34570-17-7

IUPAC Name: 2-carbamimidoylacetamide hydrochloridyl
Synonyms: 2-carbamimidoylacetamide hydrochloridyl
Molecular Weight (g/mol): 136.56
Molecular Formula: C3H7ClN3O
InChi Key: MDSJPVJGEXMXIC-UHFFFAOYSA-N
SMILES: [Cl].NC(=N)CC(N)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results