Learn More
Malonaldehyde bis(phenylimine) monohydrochloride, 97+%
CAS: 123071-42-1 | C15H15ClN2 | 258.749 g/mol
Supplier: Thermo Scientific Chemicals B2172122
Description
Malonaldehyde bis(phenylimine) monohydrochloride is a useful fluorescent cyanine dye.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMalonaldehyde bis(phenylimine) monohydrochloride is a useful fluorescent cyanine dye.
Solubility
Soluble in water (partly).
Notes
Hygroscopic, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
| Malonaldehyde bis(phenylimine) monohydrochloride | |
| ∽215°C (decomposition) | |
| MFCD00054634 | |
| malonaldehyde bis phenylimine monohydrochloride, malonaldehyde bis phenylimine hydrochloride, malondialdehyde bis phenylimine monohydrochloride, malonaldehyde bis phenylimine hcl, malonaldehydedianilidehydrochloride, n-3-phenylimino-1-propen-1-yl aniline hydrochloride, n1,n3-diphenylpropane-1,3-diimine hydrochloride, 1e,3e-n1,n3-diphenylpropane-1,3-diimine hydrochloride | |
| FLMLOZWXXPLHIE-UHFFFAOYSA-N | |
| N,N'-diphenylpropane-1,3-diimine;hydrochloride | |
| 12213307 | |
| 98% |
| 123071-42-1 | |
| C15H15ClN2 | |
| 100 g | |
| Soluble in water (partly). | |
| C1=CC=C(C=C1)N=CCC=NC2=CC=CC=C2.Cl | |
| 258.749 | |
| 258.75 |
Chemical Identifiers
| 123071-42-1 | |
| 258.749 | |
| FLMLOZWXXPLHIE-UHFFFAOYSA-N | |
| 12213307 | |
| C1=CC=C(C=C1)N=CCC=NC2=CC=CC=C2.Cl |
| C15H15ClN2 | |
| MFCD00054634 | |
| malonaldehyde bis phenylimine monohydrochloride, malonaldehyde bis phenylimine hydrochloride, malondialdehyde bis phenylimine monohydrochloride, malonaldehyde bis phenylimine hcl, malonaldehydedianilidehydrochloride, n-3-phenylimino-1-propen-1-yl aniline hydrochloride, n1,n3-diphenylpropane-1,3-diimine hydrochloride, 1e,3e-n1,n3-diphenylpropane-1,3-diimine hydrochloride | |
| N,N'-diphenylpropane-1,3-diimine;hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only