missing translation for 'onlineSavingsMsg'
Learn More

Malonaldehyde bis(phenylimine) monohydrochloride, 97+%

Catalog No. p-7025971
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2172114 25 g
AAB2172122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2172114 Supplier Thermo Scientific Chemicals Supplier No. B2172114

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 123071-42-1 | C15H15ClN2 | 258.749 g/mol

Malonaldehyde bis(phenylimine) monohydrochloride is a useful fluorescent cyanine dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Malonaldehyde bis(phenylimine) monohydrochloride is a useful fluorescent cyanine dye.

Solubility
Soluble in water (partly).

Notes
Hygroscopic, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 123071-42-1
Molecular Formula C15H15ClN2
Molecular Weight (g/mol) 258.749
MDL Number MFCD00054634
InChI Key FLMLOZWXXPLHIE-UHFFFAOYSA-N
Synonym malonaldehyde bis phenylimine monohydrochloride, malonaldehyde bis phenylimine hydrochloride, malondialdehyde bis phenylimine monohydrochloride, malonaldehyde bis phenylimine hcl, malonaldehydedianilidehydrochloride, n-3-phenylimino-1-propen-1-yl aniline hydrochloride, n1,n3-diphenylpropane-1,3-diimine hydrochloride, 1e,3e-n1,n3-diphenylpropane-1,3-diimine hydrochloride
PubChem CID 12213307
IUPAC Name N,N'-diphenylpropane-1,3-diimine;hydrochloride
SMILES C1=CC=C(C=C1)N=CCC=NC2=CC=CC=C2.Cl

Specifications

Melting Point ∽215°C (decomposition)
Quantity 25 g
Solubility Information Soluble in water (partly).
Formula Weight 258.75
Percent Purity 98%
Chemical Name or Material Malonaldehyde bis(phenylimine) monohydrochloride
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only