missing translation for 'onlineSavingsMsg'
Learn More

Malonaldehyde bis(dimethyl acetal), 98%

Catalog No. p-154813
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC148611000 100 g Glass bottle
AC148615000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC148611000 Supplier Thermo Scientific Chemicals Supplier No. 148611000
Only null left

CAS: 102-52-3 | C7H16O4 | 164.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 102-52-3
Molecular Formula C7H16O4
Molecular Weight (g/mol) 164.20
MDL Number MFCD00008488
InChI Key XHTYQFMRBQUCPX-UHFFFAOYSA-N
Synonym propane, 1,1,3,3-tetramethoxy, tetramethoxypropane, malonaldehyde bis dimethyl acetal, malonaldehyde tetramethyl acetal, malonaldehyde, bis dimethyl acetal, 1,1,3,3-tetramethoxy-propane, malonaldehyde bis dime, 1,3,3-tetramethoxypropane, ksc174o0b, propane,1,3,3-tetramethoxy
PubChem CID 66019
IUPAC Name 1,1,3,3-tetramethoxypropane
SMILES COC(CC(OC)OC)OC

Specifications

Density 0.9900g/mL
Boiling Point 183.0°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4060 to 1.4080
Linear Formula (CH3O)2CHCH2CH(OCH3)2
Quantity 100 g
Specific Gravity 0.99
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in hexane,ether,alcohol
Formula Weight 164.2
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Malonaldehyde bis(dimethyl acetal), 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 203-037-2
TSCA TSCA

RUO – Research Use Only