accessibility menu, dialog, popup

UserName

CAS RN 102-52-3

IUPAC Name: 1,1,3,3-tetramethoxypropane
Synonyms: propane, 1,1,3,3-tetramethoxy-
Molecular Weight (g/mol): 164.2
Molecular Formula: C7H16O4
InChi Key: XHTYQFMRBQUCPX-UHFFFAOYSA-N
SMILES: COC(CC(OC)OC)OC
Boiling Point 
  • (3)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (5)