missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadence Science™ Malleable Stainless Steel Animal Feeding Needles
Designed to meet the special needs of pharmacologists, toxicologists, pathologists, and other biomedical researchers working with laboratory animals. Cadence Science™ Malleable Stainless Steel Animal Feeding Needles are sterile and disposable.
Supplier: Cadence Science™ POP9917
Description
- Type 304 stainless steel construction
- Malleable
- Sterile
- Disposable
- 100/box, 20 x 5/sterile pouch
Chemical Identifiers
| Animal Feeding Needle | |
| Single-use | |
| Sterile |
| Stainless Steel (304) | |
| Animal feeding |
Specifications
| Animal Feeding Needle | |
| Single-use | |
| 18G | |
| 3.1 in. |
| Stainless Steel (304) | |
| Animal feeding | |
| Sterile | |
| 2.3 mm |