missing translation for 'onlineSavingsMsg'
Learn More

Malic acid Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015252
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015252 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015252 Supplier Merck Emd Millipore Supplier No. PHR12731G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Malic acid is a naturally occurring acidulant and low acid food additive, present in most fruits and vegetables. It is widely used to enhance the flavor, adjust the pH value of alkaline products and to control the growth of bacteria.

Specifications

CAS 6915-15-7
Ignition Point 644°F
Vapor Pressure <0.1 mmHg (20°C)
Vapor Density 4.6 (vs. air)
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula HO2 CCH2 CH(OH)CO2 H
MDL Number MFCD00064212
Melting Point 131°C to 133°C
Synonym DL-Malic acid; (+/-)-2-Hydroxysuccinic acid; DL-Hydroxybutanedioic acid
Type Certified Reference Material
Quantity 1 g
Formula Weight 134.09
Grade Certified Reference Material
Show More Show Less