missing translation for 'onlineSavingsMsg'
Learn More

Maleic Anhydride 99.0+%, TCI America™

Catalog No. p-7131920
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M000525G 25 g
M0005500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. M000525G missing translation for 'mfr' TCI America missing translation for 'supplierNo' M000525G
Only null left

Chemical Identifiers

CAS 108-31-6
Molecular Formula C4H2O3
Molecular Weight (g/mol) 98.06
MDL Number MFCD00005518
InChI Key FPYJFEHAWHCUMM-UHFFFAOYSA-N
Synonym maleic anhydride, 2,5-furandione, maleic acid anhydride, toxilic anhydride, dihydro-2,5-dioxofuran, cis-butenedioic anhydride, 2,5-dihydrofuran-2,5-dione, polymaleic anhydride, maleinanhydrid, rcra waste number u147
PubChem CID 7923
ChEBI CHEBI:474859
IUPAC Name 2,5-dihydrofuran-2,5-dione
SMILES O=C1OC(=O)C=C1

Specifications

Melting Point 54°C
Color White
Boiling Point 199°C
Quantity 25 g
UN Number 2215
Formula Weight 98.06
Percent Purity ≥99.0% (GC,T)
Physical Form Pellets
Chemical Name or Material Maleic Anhydride
EINECSNumber (2)-1101
RTECSNumber ON3675000
TSCA Yes