accessibility menu, dialog, popup

UserName

CAS RN 108-31-6

IUPAC Name: 2,5-dihydrofuran-2,5-dione
Synonyms: maleic anhydride toxilic anhydride polymaleic anhydride dihydro-2,5-dioxofuran
Molecular Weight (g/mol): 98.06
Molecular Formula: C4H2O3
InChi Key: FPYJFEHAWHCUMM-UHFFFAOYSA-N
SMILES: O=C1OC(=O)C=C1
Boiling Point 
  • (2)
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (4)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (8)
  • (2)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (4)
  • (6)
  • (4)