missing translation for 'onlineSavingsMsg'
Learn More

Maleic Anhydride 99.0+%, TCI America™

Catalog No. M0005500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0005500G 500 g
M000525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0005500G Supplier TCI America Supplier No. M0005500G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 108-31-6
Molecular Formula C4H2O3
Molecular Weight (g/mol) 98.06
MDL Number MFCD00005518
InChI Key FPYJFEHAWHCUMM-UHFFFAOYSA-N
Synonym maleic anhydride, 2,5-furandione, maleic acid anhydride, toxilic anhydride, dihydro-2,5-dioxofuran, cis-butenedioic anhydride, 2,5-dihydrofuran-2,5-dione, polymaleic anhydride, maleinanhydrid, rcra waste number u147
PubChem CID 7923
ChEBI CHEBI:474859
IUPAC Name 2,5-dihydrofuran-2,5-dione
SMILES O=C1OC(=O)C=C1

Specifications

Melting Point 54°C
Color White
Boiling Point 199°C
Quantity 500 g
UN Number 2215
Formula Weight 98.06
Percent Purity ≥99.0% (GC,T)
Physical Form Pellets
Chemical Name or Material Maleic Anhydride
EINECSNumber (2)-1101
RTECSNumber ON3675000
TSCA Yes