Learn More
Maleic anhydride, briquettes
CAS: 108-31-6 | C4H2O3 | 98.06 g/mol
$63.18 - $63.18
Chemical Identifiers
| CAS | 108-31-6 |
|---|---|
| Molecular Formula | C4H2O3 |
| Molecular Weight (g/mol) | 98.06 |
| MDL Number | MFCD00005518 |
| InChI Key | FPYJFEHAWHCUMM-UHFFFAOYSA-N |
| IUPAC Name | 2,5-dihydrofuran-2,5-dione |
| SMILES | O=C1OC(=O)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC465660010
|
Thermo Scientific Chemicals
465660010 |
1 kg |
Each for $63.18
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 108-31-6 | |
| 98.06 | |
| FPYJFEHAWHCUMM-UHFFFAOYSA-N | |
| O=C1OC(=O)C=C1 |
| C4H2O3 | |
| MFCD00005518 | |
| 2,5-dihydrofuran-2,5-dione |
Specifications
| 108-31-6 | |
| 200.0°C | |
| C4H2O3 | |
| 1 kg | |
| FPYJFEHAWHCUMM-UHFFFAOYSA-N | |
| 2,5-dihydrofuran-2,5-dione | |
| 1.6 mPa.s (60°C) | |
| Maleic anhydride |
| 52.0°C to 55.0°C | |
| 102°C | |
| MFCD00005518 | |
| Solubility aq. soln.: 400 g/L (25°C). Other solubilities: reacts with water forming maleic acid, 227 g/100 g acetone - 50 g/100 g benzene (25°C), 112 g/100 g ethylacetate (25°C)- 0,60 g/100 g ccl4, 52.5 g/100 g chloroform (25°C)-23,4 g/100 g toluen | |
| O=C1OC(=O)C=C1 | |
| 98.06 | |
| 98.06 |
Safety and Handling
GHS H Statement Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes damage to organs through
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water or shower Immediately call a POISON CENTER or doctor/physician.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Danger
EINECSNumber : 203-571-6
RUO – Research Use Only