missing translation for 'onlineSavingsMsg'
Learn More

Maleate, 0.2M buffer soln., pH 5.5, Thermo Scientific™

Catalog No. AAJ62049AP
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62049AP 500 mL
AAJ62049AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62049AP Supplier Thermo Scientific Chemicals Supplier No. J62049AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 3105-55-3
Molecular Formula C4H2NaO4
Molecular Weight (g/mol) 137.05
MDL Number MFCD00070246
InChI Key VRVKOZSIJXBAJG-ODZAUARKSA-L
PubChem CID 91658932
SMILES [Na+].[O-]C(=O)\C=C/C([O-])=O

Specifications

Color Colorless
Concentration 0.2 M
pH 5.5
Physical Form Liquid
Quantity 500 mL
IUPAC Name sodium (2Z)-but-2-enedioate
Chemical Name or Material Maleate buffer soln.
EINECSNumber 221-461-6
TSCA No
Recommended Storage Ambient temperatures