missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Maleamide, TCI America™
Supplier: TCI America M000425G
Specifications
| Maleamide | |
| White | |
| MFCD00082355 | |
| Maleic Acid Diamide | |
| NC(=O)\C=C/C(N)=O | |
| 114.10 | |
| 114.10 |
| 928-01-8 | |
| C4H6N2O2 | |
| 25 g | |
| BSSNZUFKXJJCBG-UPHRSURJSA-N | |
| (2Z)-but-2-enediamide | |
| 5355656 | |
| Crystalline Powder |
Chemical Identifiers
| 928-01-8 | |
| 114.10 | |
| BSSNZUFKXJJCBG-UPHRSURJSA-N | |
| 5355656 | |
| NC(=O)\C=C/C(N)=O |
| C4H6N2O2 | |
| MFCD00082355 | |
| Maleic Acid Diamide | |
| (2Z)-but-2-enediamide |
Safety and Handling
EINECSNumber : (2)-1121
TSCA : No