accessibility menu, dialog, popup

UserName

CAS RN 928-01-8

NH2ONH2O(Z)

CAS RN 928-01-8

IUPAC Name: (2Z)-but-2-enediamide
Synonyms: (2Z)-but-2-enediamide
Molecular Weight (g/mol): 114.1
Molecular Formula: C4H6N2O2
InChi Key: BSSNZUFKXJJCBG-UPHRSURJSA-N
SMILES: NC(=O)\C=C/C(N)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results