missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Malachite Green oxalate

CAS: 2437-29-8 | C52H54N4O12 | 927.02 g/mol

$82.81 - $192.42

Chemical Identifiers

CAS 2437-29-8
Molecular Formula C52H54N4O12
Molecular Weight (g/mol) 927.02
MDL Number MFCD00011766,MFCD00151209
InChI Key CNYGFPPAGUCRIC-UHFFFAOYSA-L
Synonym C.I. 42000
PubChem CID 2724411
IUPAC Name [4-[[4-(dimethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;2-hydroxy-2-oxoacetate;oxalic acid
SMILES OC(=O)C(O)=O.OC(=O)C([O-])=O.OC(=O)C([O-])=O.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1618614
View Documents
Thermo Scientific Chemicals
A1618614
25 g
Each for $82.81
Only null left
 
AAA1618622
View Documents
Thermo Scientific Chemicals
A1618622
100 g
Each for $192.42
Only null left
 
Description

Description

Biological stain, dye reagent, acid base indicator: pH 0.0 yellow, 2.0 green;11.6 green, 14 colorlessMalachite Green oxalate is used as the staining solution for embedded botanical material and as a spore stain. It is also used in studying the photocatalytic degradation with TiO2/WO3. It is employed for determining phosphate in serum and as a reference material in analytical chemistry for the quantification. It is also used as a pH indicator between pH 0.2 - 1.8. It is used as a dye for materials such as silk, leather, and paper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biological stain, dye reagent, acid base indicator: pH 0.0 yellow, 2.0 green;11.6 green, 14 colorlessMalachite Green oxalate is used as the staining solution for embedded botanical material and as a spore stain. It is also used in studying the photocatalytic degradation with TiO2/WO3. It is employed for determining phosphate in serum and as a reference material in analytical chemistry for the quantification. It is also used as a pH indicator between pH 0.2 - 1.8. It is used as a dye for materials such as silk, leather, and paper.

Solubility
Soluble in water, alcohol, methanol, ethanol and ethylene glycol monomethyl ether.

Notes
Incompatible with strong oxidizing agents, strong reducing agents, strong bases and strong acids.
Specifications

Chemical Identifiers

2437-29-8
927.02
CNYGFPPAGUCRIC-UHFFFAOYSA-L
2724411
OC(=O)C(O)=O.OC(=O)C([O-])=O.OC(=O)C([O-])=O.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C
C52H54N4O12
MFCD00011766,MFCD00151209
C.I. 42000
[4-[[4-(dimethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;2-hydroxy-2-oxoacetate;oxalic acid

Specifications

164°C (decomposition)
C52H54N4O12
UN2811
14,5699
Soluble in water,alcohol,methanol,ethanol and ethylene glycol monomethyl ether.
OC(=O)C(O)=O.OC(=O)C([O-])=O.OC(=O)C([O-])=O.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C
927.02
927.02
Malachite Green oxalate
2437-29-8
MFCD00011766,MFCD00151209
3644550
C.I. 42000
CNYGFPPAGUCRIC-UHFFFAOYSA-L
[4-[[4-(dimethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;2-hydroxy-2-oxoacetate;oxalic acid
2724411
25 g
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H361-H302
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.

P201-P202-P264b-P270-P281-P301+P310-P305+P351+P338-P308+P313-P310-P330-P501c

H301-H318-H361d

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 219-441-7

RTECSNumber : BQ1190000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only