Learn More
Thermo Scientific Chemicals Malachite Green oxalate, pure, certified
CAS: 2437-29-8 | C52H54N4O12 | 927.02 g/mol
$94.49 - $254.42
Chemical Identifiers
| CAS | 2437-29-8 |
|---|---|
| Molecular Formula | C52H54N4O12 |
| Molecular Weight (g/mol) | 927.02 |
| MDL Number | MFCD00011766,MFCD00151209 |
| InChI Key | CNYGFPPAGUCRIC-UHFFFAOYSA-L |
| Synonym | Basic Green 4, C.I. 42000 |
| PubChem CID | 2724411 |
| IUPAC Name | [4-[[4-(dimethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;2-hydroxy-2-oxoacetate;oxalic acid |
| SMILES | OC(=O)C(O)=O.OC(=O)C([O-])=O.OC(=O)C([O-])=O.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC413490250
|
Thermo Scientific Chemicals
413490250 |
25 g | Glass Bottle |
Each for $94.49
|
|
||||
|
AC413491000
|
Thermo Scientific Chemicals
413491000 |
100 g | Glass bottle |
Each for $254.42
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2437-29-8 | |
| 927.02 | |
| CNYGFPPAGUCRIC-UHFFFAOYSA-L | |
| 2724411 | |
| OC(=O)C(O)=O.OC(=O)C([O-])=O.OC(=O)C([O-])=O.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C |
| C52H54N4O12 | |
| MFCD00011766,MFCD00151209 | |
| Basic Green 4, C.I. 42000 | |
| [4-[[4-(dimethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;2-hydroxy-2-oxoacetate;oxalic acid |
Specifications
| 2437-29-8 | |
| 90% min. | |
| MFCD00011766,MFCD00151209 | |
| 15, 5763 | |
| CNYGFPPAGUCRIC-UHFFFAOYSA-L | |
| OC(=O)C(O)=O.OC(=O)C([O-])=O.OC(=O)C([O-])=O.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C | |
| 927.02 | |
| 614 to 620nm (in water) | |
| Pure | |
| Vapor Pressure: Negligible | |
| Green | |
| Metallic To Crystalline Powder or Crystals |
| 1.00 to 1.15 | |
| C52H54N4O12 | |
| 13, 745 | |
| Basic Green 4, C.I. 42000 | |
| Authentic | |
| [4-[[4-(dimethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;2-hydroxy-2-oxoacetate;oxalic acid | |
| 2724411 | |
| 927.03 | |
| Glass Bottle | |
| from pH 0.0 (yellow) to pH 2.0 (green) from pH 11.6 (blue-green) to pH 14.0 (colorless) | |
| 25 g | |
| Malachite Green oxalate, Pure |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Suspected of damaging the unborn child.
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.
Emergency Overview
Toxic if swallowed. Causes eye burns. Risk of serious damage to eyes. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. May cause harm to the unborn
Danger
RUO – Research Use Only