missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Malachite Green isothiocyanate

Catalog No. M689
Encompass
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M689 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M689 Supplier Invitrogen™ Supplier No. M689

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

malachite green isothiocyanate

Malachite green is a nonfluorescent photosensitizer that absorbs at long wavelengths (∼630 nm). Its photosensitizing action can be targeted to particular cellular sites by using the amine-reactive malachite green isothiocyanate to conjugate this label to antibodies or other biomolecules.

Specifications

Quantity 10 mg
Product Type Malachite Green Isothiocyanate
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Isothiocyanate
Label or Dye Malchite Green
Label Type Other Labels or Dyes
Color Green

For Research Use Only. Not for use in diagnostic procedures.