missing translation for 'onlineSavingsMsg'
Learn More

Malachite Green chloride, ≥96.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111016467
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016467 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016467 Supplier Merck Emd Millipore Supplier No. 3880025MG

Malachite green chloride is a triphenylmethane dye, which generally can find applications in aquaculture and highly effective against fungal infections.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 569-64-2
Physical Form Neat
Linear Formula C23H25 ClN2
MDL Number MFCD00011751
Synonym N,N,N′,N′-Tetramethyl-4,4 ′-diaminotriphenylcarbenium chloride; Basic Green 4; Malachite Green
Type Analytical Standard
Quantity 25 mg
Formula Weight 364.91
Percent Purity ≥96.0% (HPLC)
Grade Analytical Standard
Show More Show Less