missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MagniSort™ Streptavidin Negative Selection Beads
Designed for the magnetic separation of cells by negative selection. Undesired cells are bound by a cocktail of biotinylated antibodies and then streptavidin coated magnetic beads.
$192.00 - $710.00
Specifications
| Form | Liquid |
|---|---|
| Ligand | Streptavidin |
| Product Type | Streptavidin Conjugate (Fluorescent) |
| Target Molecule | Biotin |
Alert:
Contains Sodium azide
Description
- Clone and concentration of biotinylated antibody for undesired cell labeling, as well as concentration of MagniSort™ Streptavidin Negative Selection Beads must be determined empirically
- Please refer to the protocol for details on how to optimize both the antibody or antibody cocktail and MagniSort™ Streptavidin Negative Selection Beads.
- For depletion of cells using a single antibody, we recoomend the MagniSort™ Streptavidin Positive Selection Beads (cat. MSPB-6003) and protocol
Specifications
| Liquid | |
| Streptavidin | |
| Streptavidin Conjugate (Fluorescent) | |
| Biotin |
For Research Use Only. Not for use in diagnostic procedures.