Learn More
Synbiosis™ Magnifying Glass Adapter
Magnifying Glass Adapter to be used with aCOLade 2 colony counter for easier viewing of small colonies.
$674.58 - $976.82
Specifications
Product Type Magnifying Glass
Product Line Synbiosis
For Use With (Application) Magnification for a manual colony counter
For Use With (Equipment) aCOLade 2
View More Specs
Products 2
Catalog Number Mfr. No. Magnification Material Price Quantity  
Catalog Number Mfr. No. Magnification Material Price Quantity  
01257342
View Documents
Synbiosis™
ACOLAD2MAG1
1.7X Plastic
Each for $674.58
Only null left
01257343
View Documents
Synbiosis™
ACOLAD2MAG2
3X Glass
Each for $976.82
Only null left
Description
Description
  • Designed for use with aCOLade 2
  • Options of 1.7X (Plastic) or 3X (Glass) magnification for easier viewing of colonies
Specifications
Specifications
Magnifying Glass
Synbiosis
Magnification for a manual colony counter
aCOLade 2