Learn More
Synbiosis™ Magnifying Glass Adapter
Magnifying Glass Adapter to be used with aCOLade 2 colony counter for easier viewing of small colonies.
Supplier: Synbiosis™ ACOLAD2MAG1
Description
- Designed for use with aCOLade 2
- Options of 1.7X (Plastic) or 3X (Glass) magnification for easier viewing of colonies
Specifications
| Magnifying Glass | |
| Plastic | |
| Magnification for a manual colony counter |
| 1.7X | |
| Synbiosis | |
| aCOLade 2 |