missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Trisilicate, Powder, USP, 45%, Spectrum™ Chemical

Catalog No. p-7157797
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604140 500 g Poly Bottle
18604141 2.5 kg Poly Pail
18604142 12 kg Fiber Drum
18604143 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18604140 Supplier Spectrum Chemical Mfg Cor Supplier No. MA140500GM
Only null left

MA140, 39365-87-2, 2MgO·3SiO2·xH2O

Spectrum™ Chemical Magnesium Trisilicate, Powder, USP , often used in combination with dried aluminum hydroxide to act as an effective antacid to combat indigestion. This inorganic compound is available in it′s most common form as a powder. It is also used as a food additive in edible oils to remove impurities that occur during the frying process, it is also often employed to absorb fatty acids. Spectrum Chemical USP products, including all USP magnesium products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 39365-87-2
Molecular Formula H2MgO2Si
Molecular Weight (g/mol) 86.40
InChI Key AUDNBTJSUMKLPC-UHFFFAOYSA-N
IUPAC Name magnesium(2+) hydrate silicon oxidandiide
SMILES O.[O--].[Mg++].[Si]

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 45%
Grade USP
Packaging Poly Bottle