accessibility menu, dialog, popup

UserName

H2OOMgSi

CAS RN 39365-87-2

IUPAC Name: magnesium(2+) hydrate silicon oxidandiide
Synonyms: magnesium(2+) hydrate silicon oxidandiide
Molecular Weight (g/mol): 86.4
Molecular Formula: H2MgO2Si
InChi Key: AUDNBTJSUMKLPC-UHFFFAOYSA-N
SMILES: O.[O--].[Mg++].[Si]
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results