Learn More
Magnesium trifluoromethanesulfonate
CAS: 60871-83-2 | C2F6MgO6S2 | 322.431 g/mol
Supplier: Thermo Scientific Chemicals 02234018
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Magnesium trifluoromethanesulfonate | |
| White | |
| Powder | |
| C2F6MgO6S2 | |
| 705578 | |
| BZQRBEVTLZHKEA-UHFFFAOYSA-L | |
| magnesium;trifluoromethanesulfonate | |
| 2734125 | |
| Hygroscopic |
| >310°C | |
| 60871-83-2 | |
| 50 g | |
| MFCD00042601 | |
| magnesium trifluoromethanesulfonate, magnesium triflate, magnesium 2+ ditriflate, trifluoromethanesulfonic acid magnesium salt, magnesium trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, magnesium salt, magnesiumtrifluoromethanesulfonate, magnesium trifiate, acmc-209mmv, ksc352a4p | |
| C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Mg+2] | |
| 322.431 | |
| 322.43 |
Chemical Identifiers
| 60871-83-2 | |
| 322.431 | |
| BZQRBEVTLZHKEA-UHFFFAOYSA-L | |
| 2734125 | |
| C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Mg+2] |
| C2F6MgO6S2 | |
| MFCD00042601 | |
| magnesium trifluoromethanesulfonate, magnesium triflate, magnesium 2+ ditriflate, trifluoromethanesulfonic acid magnesium salt, magnesium trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, magnesium salt, magnesiumtrifluoromethanesulfonate, magnesium trifiate, acmc-209mmv, ksc352a4p | |
| magnesium;trifluoromethanesulfonate |
Safety and Handling
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : DOT Class: 8, Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only