missing translation for 'onlineSavingsMsg'
Learn More

Magnesium trifluoromethanesulfonate

Catalog No. AA2234009
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2234009 10 g
AA2234018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA2234009 Supplier Thermo Scientific Chemicals Supplier No. 02234009

CAS: 60871-83-2 | C2F6MgO6S2 | 322.431 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 60871-83-2
Molecular Formula C2F6MgO6S2
Molecular Weight (g/mol) 322.431
MDL Number MFCD00042601
InChI Key BZQRBEVTLZHKEA-UHFFFAOYSA-L
Synonym magnesium trifluoromethanesulfonate, magnesium triflate, magnesium 2+ ditriflate, trifluoromethanesulfonic acid magnesium salt, magnesium trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, magnesium salt, magnesiumtrifluoromethanesulfonate, magnesium trifiate, acmc-209mmv, ksc352a4p
PubChem CID 2734125
IUPAC Name magnesium;trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Mg+2]

Specifications

Melting Point >310°C
Color White
Physical Form Powder
Quantity 10 g
Beilstein 705578
Formula Weight 322.43
Sensitivity Hygroscopic
Chemical Name or Material Magnesium trifluoromethanesulfonate
Hazard Category H314
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8, Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only