missing translation for 'onlineSavingsMsg'
Learn More

Magnesium trifluoromethanesulfonate, 97%

Catalog No. p-7023737
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1759106 5 g
AAA1759114 25 g
AAA1759122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1759106 Supplier Thermo Scientific Chemicals Supplier No. A1759106

CAS: 60871-83-2 | C2F6MgO6S2 | 322.431 g/mol

Magnesium trifluoromethanesulfonate is used as a component of the electrolyte, as lewis acid resulted in largely decreased and reversed enantioselectivities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium trifluoromethanesulfonate is used as a component of the electrolyte, as lewis acid resulted in largely decreased and reversed enantioselectivities.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 60871-83-2
Molecular Formula C2F6MgO6S2
Molecular Weight (g/mol) 322.431
MDL Number MFCD00042601
InChI Key BZQRBEVTLZHKEA-UHFFFAOYSA-L
Synonym magnesium trifluoromethanesulfonate, magnesium triflate, magnesium 2+ ditriflate, trifluoromethanesulfonic acid magnesium salt, magnesium trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, magnesium salt, magnesiumtrifluoromethanesulfonate, magnesium trifiate, acmc-209mmv, ksc352a4p
PubChem CID 2734125
IUPAC Name magnesium;trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Mg+2]

Specifications

Melting Point >310°C
Linear Formula Mg(CF3SO3)2
Quantity 5 g
UN Number UN1759
Beilstein 705578
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 322.43
Percent Purity 98%
Chemical Name or Material Magnesium trifluoromethanesulfonate
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only