missing translation for 'onlineSavingsMsg'
Learn More

Magnesium tetraborate, 99%

CAS: 12007-62-4 | B2Mg3O6 | 190.529 g/mol

$225.96 - $790.86

Chemical Identifiers

CAS 12007-62-4
Molecular Formula B2Mg3O6
Molecular Weight (g/mol) 190.529
MDL Number MFCD02688001
InChI Key NFMWFGXCDDYTEG-UHFFFAOYSA-N
Synonym acmc-1bvay, boric acid h3bo3 , magnesium salt 2:3, ksc171o6l, trimagnesium 2+ diborate, parent, trimagnesium 2+ ion diborate
PubChem CID 21981398
IUPAC Name trimagnesium;diborate
SMILES B([O-])([O-])[O-].B([O-])([O-])[O-].[Mg+2].[Mg+2].[Mg+2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4389214
View Documents
Thermo Scientific Chemicals
04389214
25 g
Each for $225.96
Only null left
 
AA4389222
View Documents
Thermo Scientific Chemicals
04389222
100 g
Each for $790.86
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12007-62-4
190.529
NFMWFGXCDDYTEG-UHFFFAOYSA-N
21981398
B([O-])([O-])[O-].B([O-])([O-])[O-].[Mg+2].[Mg+2].[Mg+2]
B2Mg3O6
MFCD02688001
acmc-1bvay, boric acid h3bo3 , magnesium salt 2:3, ksc171o6l, trimagnesium 2+ diborate, parent, trimagnesium 2+ ion diborate
trimagnesium;diborate

Specifications

12007-62-4
25 g
MFCD02688001
NFMWFGXCDDYTEG-UHFFFAOYSA-N
trimagnesium;diborate
21981398
99%
Magnesium tetraborate
Powder
B2Mg3O6
acmc-1bvay, boric acid h3bo3 , magnesium salt 2:3, ksc171o6l, trimagnesium 2+ diborate, parent, trimagnesium 2+ ion diborate
B([O-])([O-])[O-].B([O-])([O-])[O-].[Mg+2].[Mg+2].[Mg+2]
190.529
179.54
Odorless
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only