accessibility menu, dialog, popup

UserName

BOMg

CAS RN 12007-62-4

IUPAC Name: magnesium(2+) boron oxidandiide
Synonyms: magnesium(2+) boron oxidandiide
Molecular Weight (g/mol): 51.11
Molecular Formula: BMgO
InChi Key: YZUWTDQUEUJLAR-UHFFFAOYSA-N
SMILES: [B].[O--].[Mg++]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results