missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Sulfate TS, 12% (w/v), Ricca Chemical
$113.19 - $113.19
Chemical Identifiers
| CAS | 10034-99-8 |
|---|---|
| Molecular Formula | H14MgO11S |
| Molecular Weight (g/mol) | 246.47 |
| MDL Number | MFCD00149785 |
| InChI Key | WRUGWIBCXHJTDG-UHFFFAOYSA-L |
| Synonym | magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate |
| PubChem CID | 24843 |
| ChEBI | CHEBI:31795 |
| IUPAC Name | magnesium(2+) heptahydrate sulfate |
| SMILES | O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O |
Chemical Identifiers
| 10034-99-8 | |
| 246.47 | |
| WRUGWIBCXHJTDG-UHFFFAOYSA-L | |
| 24843 | |
| magnesium(2+) heptahydrate sulfate |
| H14MgO11S | |
| MFCD00149785 | |
| magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate | |
| CHEBI:31795 | |
| O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O |
Specifications
| Colorless | |
| 87.0,10.51 | |
| H14MgO11S | |
| magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate | |
| O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O | |
| 246.47 | |
| CHEBI:31795 | |
| Laboratory | |
| Aqueous Solution | |
| Liquid | |
| Magnesium Sulfate, Test Solution |
| 10034-99-8,7732-18-5 | |
| 91.0,10.93 | |
| MFCD00149785 | |
| WRUGWIBCXHJTDG-UHFFFAOYSA-L | |
| magnesium(2+) heptahydrate sulfate | |
| 24843 | |
| 12% | |
| Natural Poly Bottle | |
| 500 mL | |
| 12% (w/v) |