missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Sulfate Solution, 250 g/L in 2.0 N Sulfuric Acid, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | MAGNESIUM SULFATE SOLUTION |
| CAS | 7487-88-9, 10034-99-8, 7664-93-9 |
| CAS Min % | 73.0, 18.14, 7.34 |
| CAS Max % | 75.0, 18.88, 7.64 |
| Synonym | dihydrogen oxide, dihydrogen monoxide |
| Grade | EP |
| Packaging | Natural Poly Bottle |
| pH | <1 |
| Quantity | 500 mL |