Learn More
Magnesium Sulfate, Anhydrous, USP, 99-100.5%, Spectrum™ Chemical
M1354, 7487-88-9, MgSO4
Supplier:  Spectrum Chemical Mfg Cor M135412KG
 View more versions of this product
Catalog No. 18604128
Only null left
Explore available promotions
Description
Description
Spectrum™ Chemical Magnesium Sulfate, Anhydrous, USP is the raw material for a common magnesium-based, pharmaceutical preparation called Epsom salts. The Epsom salt formulations are for both topical and oral indications. The topical preparation is a gel for soothing aches and pains while the internal formulation is commonly used as saline laxative. In it′s anhydrous form, magnesium sulfate is used as a drying agent. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities..
Specifications
Specifications
7487-88-9
5.0-9.2
MgO4S
CSNNHWWHGAXBCP-UHFFFAOYSA-L
magnesium(2+) sulfate
99 to 100.5%
2%
100%
12 kg
MFCD00011110
[Mg++].[O-]S([O-])(=O)=O
120.36
USP
Poly Pail
Chemical Identifiers
7487-88-9
120.36
CSNNHWWHGAXBCP-UHFFFAOYSA-L
[Mg++].[O-]S([O-])(=O)=O
MgO4S
MFCD00011110
magnesium(2+) sulfate
Product Suggestions
Product Suggestions