missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate, Anhydrous, USP, 99-100.5%, Spectrum™ Chemical

Catalog No. p-7157749
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604125 125 g Poly Bottle
18604126 500 g Poly Bottle
18604127 2.5 kg Poly Pail
18604130 5 kg Poly Pail
18604128 12 kg Poly Pail
18604129 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18604125 Supplier Spectrum Chemical Mfg Cor Supplier No. M1354125GM
Only null left

M1354, 7487-88-9, MgSO4

Spectrum™ Chemical Magnesium Sulfate, Anhydrous, USP is the raw material for a common magnesium-based, pharmaceutical preparation called Epsom salts. The Epsom salt formulations are for both topical and oral indications. The topical preparation is a gel for soothing aches and pains while the internal formulation is commonly used as saline laxative. In it′s anhydrous form, magnesium sulfate is used as a drying agent. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities..

Chemical Identifiers

CAS 7487-88-9
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.36
MDL Number MFCD00011110
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
IUPAC Name magnesium(2+) sulfate
SMILES [Mg++].[O-]S([O-])(=O)=O

Specifications

pH 5.0-9.2
CAS Max % 100%
Quantity 125 g
Percent Purity 99 to 100.5%
Grade USP
Loss on Drying 2%
Packaging Poly Bottle