missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate, Anhydrous, Powder, CP, Spectrum™ Chemical

M1064, 7487-88-9, MgSO4

Supplier:  Spectrum Chemical Mfg Cor M106412KG

 View more versions of this product

Catalog No. 18604123


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Magnesium Sulfate, Anhydrous, Powder, CP is hygroscopic and used as a drying agent. CP or Chemically Pure grade products supplied by Spectrum are indicative of a grade suitable for general industrial use. These materials may or may not have a Certificate of Analysis available.
Specifications

Specifications

7487-88-9
12 kg
MFCD00011110
[Mg++].[O-]S([O-])(=O)=O
120.36
Poly Pail
100%
MgO4S
CSNNHWWHGAXBCP-UHFFFAOYSA-L
magnesium(2+) sulfate
Chemically Pure

Chemical Identifiers

7487-88-9
120.36
CSNNHWWHGAXBCP-UHFFFAOYSA-L
[Mg++].[O-]S([O-])(=O)=O
MgO4S
MFCD00011110
magnesium(2+) sulfate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions