missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate, Anhydrous, Powder, CP, Spectrum™ Chemical

Catalog No. p-7157698
Change view
Click to view available options
Quantity:
110 lb.
12 kg
2.5 kg
500 g
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18604121 500 g Poly Bottle
18604124 110 lb. Fiber Drum
18604122 2.5 kg Poly Pail
18604123 12 kg Poly Pail
4 options

Catalog No. 18604121

Supplier: Spectrum Chemical Mfg Cor M1064500GM

Only null left

M1064, 7487-88-9, MgSO4

Spectrum™ Chemical Magnesium Sulfate, Anhydrous, Powder, CP is hygroscopic and used as a drying agent. CP or Chemically Pure grade products supplied by Spectrum are indicative of a grade suitable for general industrial use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 7487-88-9
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.36
MDL Number MFCD00011110
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
IUPAC Name magnesium(2+) sulfate
SMILES [Mg++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 500 g
Grade Chemically Pure
Packaging Poly Bottle