missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Stearate, NF, BP, JP, 4-5%, Spectrum™ Chemical
MA130, 557-04-0
Supplier: Spectrum Chemical Mfg Cor MA13025KGCS4
Description
Spectrum™ Chemical Magnesium Stearate, NF, BP, JP is used as an anti-adherent in the manufacture of of medical capsules, tablets, and powders. Spectrum Chemical manufactured NF, BP, JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 557-04-0 | |
| 4 x 2.5 kg | |
| HQKMJHAJHXVSDF-UHFFFAOYSA-L | |
| magnesium(2+) dioctadecanoate | |
| 4 to 5% | |
| Poly Pail |
| 100% | |
| C36H70MgO4 | |
| [Mg++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O | |
| 591.26 | |
| 6% |
Chemical Identifiers
| 557-04-0 | |
| 591.26 | |
| magnesium(2+) dioctadecanoate |
| C36H70MgO4 | |
| HQKMJHAJHXVSDF-UHFFFAOYSA-L | |
| [Mg++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O |